EMAIL THIS PAGE TO A FRIEND
Journal of virology
The Src family kinase c-Yes is required for maturation of West Nile virus particles.
PMID 16140770
Abstract
The role of cellular genes in West Nile virus (WNV) replication is not well understood. Examination of cellular transcripts upregulated during WNV infection revealed an increase in the expression of the src family kinase (SFK) c-Yes. WNV-infected cell lines treated with the SFK inhibitor PP2 demonstrated a 2- to 4-log decrease in viral titers, suggesting that SFK activity is required for completion of the viral replication cycle. RNA interference mediated knock-down of c-Yes, but not c-Src, and similarly reduced virus yield, specifically implicating c-Yes in WNV production. Interestingly, PP2 treatment did not reduce intracellular levels of either viral RNA or protein, suggesting that the drug does not act on the early stages of replication. However, endoglycosidase H (endoH) digestion of the viral envelope (E) glycoprotein revealed that the acquisition of endoH-resistant glycans by E, but not endogenous major histocompatibility complex class I, was reduced in PP2-treated cells, demonstrating that E specifically does not traffic beyond the endoplasmic reticulum in the absence of SFK activity. Electron microscopy further revealed that PP2-treated WNV-infected cells accumulated an increased number of virions in the ER compared to untreated cells. Therefore, we conclude that inhibition of SFK activity did not interfere with virus assembly but prevented transit of virions through the secretory pathway. These results identify c-Yes as a cellular protein that is involved in WNV assembly and egress.