EMAIL THIS PAGE TO A FRIEND
Electrophoresis
Study on sensitivity development of 2,3-dihydroxybenzoic acid by capillary zone electrophoresis-amperometric detection with p-methyl benzoate as stacking agent.
PMID 16143980
Abstract
For its high reactivity and very short half-life, the hydroxyl radical (OH.) in vivo is very difficult to be detected. Usually, it is indirectly quantified by determining 2,3-dihydroxybenzoic acid (2,3-DHBA) and 2,5-dihydroxybenzoic acid (2,5-DHBA), which are the reaction products of salicylic acid (SA) and OH.. Because 2,5-DHBA could be directly formed by the P(450) enzyme, only 2,3-DHBA is regarded as the real biomarker of OH.in biological studies. But the very low concentration of OH* in human bodies makes its determination very difficult and complicated. In this paper, a simple online stacking capillary zone electrophoresis coupled with amperometric detection (CZE-AD) method was explored to improve the detection sensitivity of 2,3-DHBA to reach the requirements in biological analysis. A mixture solution of 12.5 mmol/L Na(2)B(4)O(7)-25 mmol/L NaH(2)PO(4) (pH 7.9) was used as the running buffer and p-methyl benzoate was selected as a suitable stacker. The effects of the concentration, pH value, and injection time of p-methyl benzoate on stacking efficiency were carefully studied. Under the optimum stacking CZE-AD conditions, the detection sensitivity of 2,3-DHBA was improved about 20-fold and its detection limit reached the 10(-9) mol/L level. The experimental results showed that this was a potential method to determine OH* in vivo.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T36803
p-Toluic acid, 98%
C8H8O2