EMAIL THIS PAGE TO A FRIEND
Science (New York, N.Y.)
Immunology. Dietary factors and immunological consequences.
PMID 16150998
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T6330
THI
C9H14N2O5