EMAIL THIS PAGE TO A FRIEND
Biomedical research (Tokyo, Japan)
Hydroxyl radical scavenging effects of guaiacol used in traditional dental pulp sedation: reaction kinetic study.
PMID 16152729
Abstract
Guaiacol, which is a phenolic compound with a methoxy group and used in traditional dental pulp sedation, has the property of inducing cell proliferation. To clarify these mechanisms of guaiacol, this study examined the hydroxyl radical (*OH) scavenging effects of guaiacol in vitro. Generation of *OH was carried out by the Fenton reaction using mixture of ascorbic acid, H2O2, and Fe(III)-EDTA, and *OH was detected by measuring the *OH-mediated production of degradation products of deoxyribose, which reacts with 2-thiobarbituric acid (TBA) and is relatively stable for a long time. At concentrations of 10(-10) M to 10(-3) M, guaiacol inhibited the TBA reactive substance (TBA-RS) formation in a dose-dependent manner. Phenol and formaldehyde were also found to inhibit the TBA-RS formation, but their inhibitory activities were lower than that of guaiacol. The concentrations of guaiacol, phenol, and formaldehyde needed to cause 50% inhibition of TBA-RS formation were approximately 5 x 10(-6), 5 x 10(-5), and 2 x 10(-3) M, respectively. In this reaction system, guaiacol showed no chelating reaction with ferrous ion and did not directly react with H2O2. Guaiacol also exhibited radical scavenging activity on 1,1-diphenyl-2-picrylhydrazyl (DPPH) stable free radical, but its scavenging activity was lower than that toward *OH. These results suggest that guaiacol is a potent scavenger of reactive oxygen radicals and that its radical scavenging activity may be associated with its effect on cell proliferation.