EMAIL THIS PAGE TO A FRIEND
Biomacromolecules
Structural transformation of apocytochrome c induced by alternating copolymers of maleic acid and alkene.
PMID 16153115
Abstract
Apocytochrome c interacts with two copolymers: poly(isobutylene-alt-maleic acid) (PIMA) and poly(1-tetradecene-alt-maleic acid) (PTMA). The interaction leads to apocytochrome c, a conformational change from random coil to alpha-helical structure. The alpha-helix content is influenced by the copolymer concentration, the length of alkyl chain of the copolymers, and pH of the medium. The electrostatic attraction between the copolymer and protein is an indispensable factor for the folding of the protein at acid pH. The hydrophobic interaction is an important factor over the entire pH range, especially when both the copolymer and protein carry negative charges at alkaline pH. The electrostatic and hydrophobic attractions between the copolymer and protein exclude water molecules, promoting the formation of hydrogen bonds within the helical structure. On the other hand, the hydrogen bonds formed between the ionized carboxyl of the copolymer and the amide of the protein partly restrain the formation of hydrogen bonds within the helical structure when the copolymer concentration is higher at pH 6.5 and 10.5.