EMAIL THIS PAGE TO A FRIEND
Annals of the New York Academy of Sciences
Characterization of enzymatically synthesized diferulate.
PMID 16154979
Abstract
Horseradish peroxidase was used to synthesize diferulates by a procedure in which ethyl ferulate was used as substrate. Four different forms were obtained, of which two dominant were the 5-5' and 8-5' diferulate. Fluorescence emission spectra of the diferulates (excited at 284 nm) indicate that they contain two chromophores, as opposed to the substrate molecule. Fluorescence excitation spectra with emission at 417 nm further demonstrate the difference between the synthesized diferulates and starting substrates.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
320617
Ethyl 4-hydroxy-3-methoxycinnamate, 98%
C12H14O4