EMAIL THIS PAGE TO A FRIEND
Journal of economic entomology
Selective insecticide-induced stimulation on fecundity and biochemical changes in Tryporyza incertulas (Lepidoptera: Pyralidae).
PMID 16156564
Abstract
The use of selective insecticides in rice, Oryza sativa L., fields often causes resurgence of nontarget pest insects. This study was conducted to investigate the effect of two selective insecticides, buprofezin and imidacloprid, on Tryporyza incertulas (Walker), a nontarget pest. After larval feeding on rice plants treated with each insecticide, fecundity, ovary protein content, and titer of juvenile hormone III (JHIII) in the resulting female moths were determined with 'Xiushui 63' rice susceptible to T. incertulas and 'Zhendao 2' moderately resistant to T. incertulas. The fecundity of females developed from larvae that fed on the insecticide-treated Xiushui 63 plants was stimulated compared with that of moths from larvae that fed on rice plants that were not treated with either insecticide. There was no stimulating effect in females from larvae that fed on insecticide-treated Zhendao 2 plants. The weight of fourth instars (final instars) that fed on the insecticide-treated Xiushui 63 rice plants was significantly greater than that of control, increasing by 50.3 and 46.7% for 60 and 112.5 g (AI) ha(-1) buprofezin, and by 23.7 and 19.5% for 15 and 37.5 g (AI) ha(-1) imidacloprid treatments, respectively. Ovary protein content in adult females developed from larvae that fed on the rice treated with the high dose of buprofezin was significantly higher than that in control. For the high and low doses of imidacloprid during the second instar, and the low dose of imidacloprid during the fourth instar, JHIII titers in female adults were also significantly higher than that in control, increasing by 152.81, 90.52, and 114.19%, respectively.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
37886
Buprofezin, PESTANAL®, analytical standard
C16H23N3OS