EMAIL THIS PAGE TO A FRIEND
Analytical and bioanalytical chemistry
Real-time monitoring of L-glutamate release from mouse brain slices under ischemia with a glass capillary-based enzyme electrode.
PMID 16158299
Abstract
Real-time monitoring of L-glutamate release from various neuronal regions of mouse hippocampal slices under ischemia (a glucose-free hypoxia condition) is described. A glass capillary microelectrode with a tip size of approximately 10 microm containing a very small volume ( approximately 2 microL) of a solution of glutamate oxidase (GluOx) and ascorbate oxidase was used. First, the amperometric response behavior of the electrode at 0 V versus Ag/AgCl was characterized with a standard glutamate solution in terms of continuous measurements, effect of oxygen, viscosity of solution and concentration dependence. The electrode was applied to the real-time monitoring of L-glutamate released from different neuronal regions of acute hippocampal slices submerged in a hypoxia solution. The time-resolved amounts of L-glutamate released at various neuronal regions (CA1, CA3 and DG) of mouse hippocampal slices were quantified and compared with the reported L-glutamate fluxes using difference-image analysis during ischemia.