EMAIL THIS PAGE TO A FRIEND
Journal of applied microbiology
Cationic antiseptics: diversity of action under a common epithet.
PMID 16162221
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A8986
Alexidine dihydrochloride, ≥95% (HPLC)
C26H56N10 · 2HCl