EMAIL THIS PAGE TO A FRIEND
Environmental research
Effects of fentin hydroxide on reproduction of the Japanese quail (Coturnix coturnix japonica).
PMID 16162336
Abstract
In a one-generation reproductive study, the fungicidal compound triphenyltin hydroxide (fentin) was administered to adult Japanese quail for 6 weeks at dietary levels of 3 and 30 ppm. Reproduction was significantly impaired in the high-dose group. The principal adverse finding was a marked increase in embryonic mortality, resulting in a lower hatch rate. Furthermore, a reduction in egg production was observed with ongoing duration of treatment. Most of the other reproduction-related parameters were not affected. The in ovo losses are assumed to result from a direct toxic effect of the test substance on chick embryos. At the low dietary level, reproduction was not altered. In contrast to the obvious reproductive toxicity, there was only limited evidence of adverse treatment-related findings in the adult birds. However, because such minor effects as an increase in mean liver weight, which was accompanied by macroscopic liver findings and a decrease in T4 serum concentration, were still seen at 3 ppm, a no-observed-effect level could not be established.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
45493
Fentin hydroxide, PESTANAL®, analytical standard
C18H16OSn
348104
Triphenyltin hydroxide
C18H16OSn