EMAIL THIS PAGE TO A FRIEND
FEMS microbiology letters
Photoactivated perylenequinone toxins in fungal pathogenesis of plants.
PMID 16165316
Abstract
Several genera of plant pathogenic fungi produce photoactivated perylenequinone toxins involved in pathogenesis of their hosts. These toxins are photosensitizers, absorbing light energy and generating reactive oxygen species that damage the membranes of the host cells. Studies with toxin-deficient mutants and on the involvement of light in symptom development have documented the importance of these toxins in successful pathogenesis of plants. This review focuses on the well studied perylenequinone toxin, cercosporin, produced by species in the genus Cercospora. Significant progress has been made recently on the biosynthetic pathway of cercosporin, with the characterization of genes encoding a polyketide synthase and a major facilitator superfamily transporter, representing the first and last steps of the biosynthetic pathway, as well as important regulatory genes. In addition, the resistance of Cercospora fungi to cercosporin and to the singlet oxygen that it generates has led to the use of these fungi as models for understanding cellular resistance to photosensitizers and singlet oxygen. These studies have shown that resistance is complex, and have documented a role for transporters, transient reductive detoxification, and quenchers in cercosporin resistance.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C6696
Cercosporin from Cercospora hayii
C29H26O10