EMAIL THIS PAGE TO A FRIEND
Biochemical and biophysical research communications
Survivin inhibits anti-growth effect of p53 activated by aurora B.
PMID 16171786
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
06-755
Anti-Histone H3 Antibody, clone, 0.5 mg/mL, Upstate®