EMAIL THIS PAGE TO A FRIEND
Pest management science
Pencycuron dissipation in soil: effect of application rate and soil conditions.
PMID 16187259
Abstract
Dissipation of the fungicide pencycuron was examined under controlled laboratory conditions in clay loam soils from rice cultivated fields of alluvial soil (Typic udifluvent) and coastal saline soil (Typic endoaquept) at field rate, twice the field rate and 10 times the field rate with and without decomposed cow manure maintained at 60% of maximum water-holding capacity (WHC) and waterlogged soil condition. The half-life values depended on the initial concentrations of pencycuron. Pencycuron, in general, degraded fastest in coastal saline soil and in soil amended with decomposed cow manure at 60% of maximum WHC of soil.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
31118
Pencycuron, PESTANAL®, analytical standard
C19H21ClN2O