EMAIL THIS PAGE TO A FRIEND
Ultrasonics sonochemistry
Sonochemical cycloadditions in ionic liquids. Lessons from model cases involving common dienes and carbonyl dienophiles.
PMID 16188479
Abstract
This contribution describes a series of sonochemical cycloadditions involving either cyclopentadiene or 1,3-cyclohexadiene with carbonyl dienophiles in an imidazolium-based ionic liquid as reaction medium. In general, ultrasound does effectively improve these processes in terms of higher yields and/or shorter reaction times when compared with the corresponding silent reactions. Stereoselectivities, however, remain practically unaffected by sonication. The role of ionic liquids under ultrasonic activation is also discussed.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C100005
1,3-Cyclohexadiene, contains 0.05% BHT as inhibitor, 97%
C6H8