EMAIL THIS PAGE TO A FRIEND
The European journal of neuroscience
Abnormal neurodevelopment, neurosignaling and behaviour in Npas3-deficient mice.
PMID 16190882
Abstract
Npas3 is a member of the bHLH-PAS superfamily of transcription factors that is expressed broadly in the developing neuroepithelium. To study the function of this gene, mice deficient in Npas3 were generated and characterized. Npas3-/- mice were growth-retarded and exhibited developmental brain abnormalities that included a reduction in size of the anterior hippocampus, hypoplasia of the corpus callosum and enlargement of the ventricles. A number of behavioural abnormalities were identified in Npas3-/- mice including locomotor hyperactivity, subtle gait defects, impairment of prepulse inhibition of acoustic startle, deficit in recognition memory and altered anxiety-related responses. Characterization of neurosignaling pathways using several pharmacological agents revealed dysfunctional glutamate, dopamine and serotonin neurotransmitter signaling. Consistent with these findings, we identified a significant alteration in cortical PSD-95 expression, a PDZ-containing protein that has been shown to be involved in postsynaptic signal transduction. Together, our observations indicate an important role for Npas3 in controlling normal brain development and neurosignaling pathways.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB3500962
Anti-NPAS3 antibody produced in rabbit, affinity isolated antibody