EMAIL THIS PAGE TO A FRIEND
Acta poloniae pharmaceutica
Applications of derivative UV spectrophotometry for the determinations of fenbufen and ketoprofen in pharmaceutical preparations.
PMID 16193808
Abstract
A methods for the determination of fenbufen and ketoprofen in pharmaceuticals by classical spectrophotometry--zero order derivative, first and second order derivatives spectrophotometry is described. using "peak-peak" (P-P) and, "peak-zero" (P-O) measurements. The calibration curves are linear within the concentration range of 2.0 - 12.0 microg x mL(-1) for fenbufen and ketoprofen. The procedure is simple, rapid and the results are reliable.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F8755
Fenbufen, analytical standard
C16H14O3
538515
Fenbufen, 96%
C16H14O3