EMAIL THIS PAGE TO A FRIEND
The Journal of laboratory and clinical medicine
Inhibitory effects of lycopene on in vitro platelet activation and in vivo prevention of thrombus formation.
PMID 16194683
Abstract
Lycopene is a natural carotenoid antioxidant that is present in tomatoes and tomato products. The pharmacologic function of lycopene in platelets is not yet understood. Therefore, in this study we sought to systematically examine the effects of lycopene in the prevention of platelet aggregation and thrombus formation. We found that lycopene concentration-dependently (2-12 micromol/L) inhibited platelet aggregation in human platelets stimulated by agonists. Lycopene (6 and 12 micromol/L) inhibited phosphoinositide breakdown in platelets labeled with tritiated inositol, intracellular Ca+2 mobilization in Fura-2 AM-loaded platelets, and thromboxane B2 formation stimulated by collagen. In addition, lycopene (6 and 12 micromol/L) significantly increased the formations of cyclic GMP and nitrate but not cyclic AMP in human platelets. Rapid phosphorylation of a protein of 47,000 Da (P47), a marker of protein kinase C activation, was triggered by PDBu (60 nmol/L). This phosphorylation was markedly inhibited by lycopene (12 micromol/L) in phosphorus-32-labeled platelets. In an in vivo study, thrombus formation was induced by irradiation of mesenteric venules in mice pretreated with fluorescein sodium. Lycopene (5, 10, and 20 mg/kg) significantly prolonged the latency period for the induction of platelet-plug formation in mesenteric venules. These results indicate that the antiplatelet activity of lycopene may involve the following pathways: (1) Lycopene may inhibit the activation of phospholipase C, followed by inhibition of phosphoinositide breakdown and thromboxane B2 formation, thereby leading to inhibition of intracellular Ca+2 mobilization. (2) Lycopene also activated the formations of cyclic GMP/nitrate in human platelets, resulting in the inhibition of platelet aggregation. The results may imply that tomato-based foods are especially beneficial in the prevention of platelet aggregation and thrombosis.