EMAIL THIS PAGE TO A FRIEND
Journal of environmental science and health. Part. B, Pesticides, food contaminants, and agricultural wastes
Fate of mutagenicity produced during anaerobic biodegradation of the herbicide chlornitrofen (CNP).
PMID 16194922
Abstract
The mutagenicity of chlornitrofen (CNP)-containing solutions has been reported to increase during anaerobic biodegradation. In the present study, the fate of this increased mutagenicity under subsequent aerobic and anaerobic incubation conditions was investigated using two Salmonella tester strains, YG 1024 (a frameshift-detecting strain) and YG 1029 (a base-pair-substitution-detecting strain). Mutagenicity for both YG 1024 and YG 1029 strains increased during nine-day anaerobic biodegradation. During subsequent anaerobic incubation, the increased mutagenicity decreased gradually for YG 1029 but did not change significantly for YG 1024. By contrast, the increased mutagenicity decreased rapidly after the conversion to aerobic incubation for both YG 1024 and YG 1029 strains. The rapid decrease in mutagenicity during aerobic incubation was due to decreases, not only in an identified mutagenic metabolite (CNP-amino) but also in unidentified mutagenic metabolites.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
04287
Chlornitrofen, PESTANAL®, analytical standard
C12H6Cl3N1O3