EMAIL THIS PAGE TO A FRIEND
Environmental health perspectives
Lead contamination in cocoa and cocoa products: isotopic evidence of global contamination.
PMID 16203244
Abstract
In this article we present lead concentrations and isotopic compositions from analyses of cocoa beans, their shells, and soils from six Nigerian cocoa farms, and analyses of manufactured cocoa and chocolate products. The average lead concentration of cocoa beans was