EMAIL THIS PAGE TO A FRIEND
Progress in clinical and biological research
Relevance of quantitation of benzo(a)pyrene metabolites in animal excretes to evaluate individual human cancer risk.
PMID 1620717