EMAIL THIS PAGE TO A FRIEND
Organic letters
Regioselective synthesis of trisubstituted 2,3-dihydrofurans from donor-acceptor cyclopropanes or from reaction of the Corey ylide with alpha-sulfenyl-, alpha-sulfinyl-, or alpha-sulfonylenones.
PMID 16209480
Abstract
[reactions: see text] Regioselective synthesis of 2,4,5- or 3,4,5-trisubstituted 2,3-dihydrofurans has been realized by using donor-acceptor cyclopropanes or by a Corey ylide reaction with alpha-sulfenyl-, alpha-sulfinyl-, or alpha-sulfonylenones. The method allowed a straightforward synthesis of the natural product calyxolane B.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
200018
2,3-Dihydrofuran, 99%
C4H6O