EMAIL THIS PAGE TO A FRIEND
Organic letters
A practical synthesis of (+/-)-alpha-isosparteine from a tetraoxobispidine core.
PMID 16209519
Abstract
[reaction: see text] The title alkaloid was synthesized in racemic form from 3,7-diallyl-2,4,6,8-tetraoxo-3,7-diazabicyclo[3.3.1]nonane (7) by a regioselective diallylation reaction followed by double ring-closing olefin metathesis and exhaustive reduction. Tetraoxobispidine 7 was itself prepared in three simple operations from dimethyl malonate. The entire sequence to alpha-isosparteine was conducted on a multigram scale and proceeded without recourse to chromatography.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
136441
Dimethyl malonate, 98%
C5H8O4
63380
Dimethyl malonate, purum, ≥96.0% (GC)
C5H8O4