EMAIL THIS PAGE TO A FRIEND
The Journal of organic chemistry
Stereoselective formation of carbon-carbon bonds via SN2-displacement: synthesis of substituted cycloalkyl[b]indoles.
PMID 16209582
Abstract
A general asymmetric synthesis of substituted cycloalkyl[b]indoles has been accomplished. The key features of this approach are (1) the utilization of a Japp-Klingemann condensation/Fischer cyclization to prepare cycloalkyl[b]indolones, (2) the asymmetric borane reduction of these heterocyclic ketones with (S)-OAB to obtain enantiomerically pure alcohols, and (3) the stereoselective S(N)2-displacement of these indole alcohol substrates with a carbon nucleophile under Mitsunobu conditions to set the C1 or C3 tertiary carbon stereocenter. The use of trimethylphosphine (PMe3) and bis(2,2,2-trichloroethyl) azodicarboxylate (TCEAD) was found to have an effect on the Mitsunobu dehydrative alkylation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
291536
Bis(2,2,2-trichloroethyl) azodicarboxylate, ≥97%
C6H4Cl6N2O4