EMAIL THIS PAGE TO A FRIEND
Chemical record (New York, N.Y.)
Electronically-coupled MM quadruply-bonded complexes of molybdenum and tungsten.
PMID 16211587
Abstract
The reaction of M2(O2CBu(t))4 (M = Mo, W) with a dicarboxylic acid in toluene yields compounds of general formula [M2]-O2C-X-CO2-[M2] ([M2] = M2(O2CBu(t))3; X = conjugated spacer). The M2 units are electronically coupled via interactions between the M2 delta and dicarboxylate pi* orbitals, and the magnitude of this coupling is revealed by electronic structure calculations and spectroscopic data. These compounds show intense metal to ligand charge transfer (MLCT) absorptions in the visible region of the electronic spectrum that are temperature and solvent dependent. Evidence of electronic coupling is seen in their cyclic voltammograms, which show two successive one-electron oxidations. The extent of electronic coupling in the mixed valence radical cations [M2]-O2C-X-CO2-[M2]+, generated by oxidation with one equivalent of AgPF6 or FeCp2PF6, is evaluated by EPR and UV-vis-NIR spectroscopic data, and delocalized behavior is observed in compounds with W2 units separated by up to 13.6 angstroms. The simplicity of the frontier M2 orbital interactions with the bridge pi orbitals provides a convenient system with which to study electron transfer in mixed valence systems, as compared to the extensively studied, but more complicated, dinuclear t(2g)6/t(2g)5 mixed valence compounds. Oligomeric and polymeric compounds incorporating M2 units have also been synthesized, having general formula [M2(O2CR)2(O2C-Thio-CO2)]n (Thio = n-hexyl substituted ter- and quinque-thiophenes). They can be deposited as thin films by spin coating, and show photoluminescence and electroluminescence. These metallo-polythiophenes show potential for application in electronic materials. (
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
767522
Tungsten, sputtering target, diam. × thickness 2.00 in. × 0.25 in., 99.95% trace metals basis
W
GF12269998
Tungsten, wire reel, 10m, diameter 0.08mm, clean, 99.95%
W
GF81410509
Tungsten, wire reel, 10m, diameter 0.08mm, gold plated, 99.95%
W
GF09514830
Tungsten, wire reel, 10m, diameter 0.125mm, clean, 99.95%
W
GF02629571
Tungsten, foil, 0.5m coil, thickness 0.035mm, coil width .5mm, as rolled, 99.95%
W
GF35593555
Tungsten, foil, 0.5m coil, thickness 0.05mm, coil width 2mm, as rolled, 99.95%
W
GF90049574
Tungsten, foil, 0.5m coil, thickness 0.2mm, coil width .6mm, as rolled, 99.95%
W
GF83538490
Tungsten, foil, 1m coil, thickness 0.02mm, coil width .1mm, as rolled, 99.95%
W
GF60432207
Tungsten, foil, 1m coil, thickness 0.05mm, coil width .51mm, as rolled, 99.95%
W
GF91197748
Tungsten, foil, 1m coil, thickness 0.1mm, coil width .6mm, as rolled, 99.95%
W
GF99741741
Tungsten, foil, 2m coil, thickness 0.025mm, coil width .7mm, as rolled, 99.95%
W
GF66018999
Tungsten, foil, 2m coil, thickness 0.04mm, coil width 0.7mm, as rolled, 99.95%
W
GF17648589
Tungsten, foil, 2m coil, thickness 0.1mm, coil width .6mm, as rolled, 99.95%
W
GF18728961
Tungsten, microfoil, disks, 25mm, thinness 0.1μm, specific density 182μg/cm2, permanent mylar 3.5μm support, 99.95%
W
GF22706210
Tungsten, rod, 1000mm, diameter 5.0mm, centerless ground, 99.95%
W
GF01295502
Tungsten, rod, 100mm, diameter 20.0mm, centerless ground, 99.95%
W
GF30443554
Tungsten, rod, 18mm, diameter 20.0mm, centerless ground, 99.95%
W
GF33990273
Tungsten, rod, 200mm, diameter 3.0mm, centerless ground, 99.95%
W
GF38415023
Tungsten, rod, 500mm, diameter 10.0mm, centerless ground, 99.95%
W
GF21910700
Tungsten, rod, 500mm, diameter 8.0mm, centerless ground, 99.95%
W
GF14172612
Tungsten, wire reel, 0.5m, diameter 0.25mm, clean, 99.95%
W
GF79162645
Tungsten, wire reel, 1000m, diameter 0.04mm, clean, 99.95%
W
GF15895097
Tungsten, wire reel, 25m, diameter 0.005mm, gold plated, 99.95%
W
GF87362118
Tungsten, wire reel, 25m, diameter 0.015mm, gold plated, 99.95%
W
GF60349989
Tungsten, wire reel, 25m, diameter 0.01mm, gold plated, 99.95%
W
GF46434281
Tungsten, wire reel, 25m, diameter 0.025mm, clean, 99.95%
W
GF27705281
Tungsten, wire reel, 25m, diameter 0.02mm, gold plated, 99.95%
W
GF94019586
Tungsten, wire reel, 25m, diameter 0.030mm, gold plated, 99.95%
W
GF48200576
Tungsten, wire reel, 25m, diameter 0.035mm, clean, 99.95%
W
GF07740682
Tungsten, wire reel, 25m, diameter 0.04mm, clean, 99.95%
W