EMAIL THIS PAGE TO A FRIEND
Microbial ecology
Microcystins induce morphological and physiological changes in selected representative phytoplanktons.
PMID 16215645
Abstract
Dissolved microcystins (MC) are regularly present in water dominated by microcystin-producing, bloom-forming cyanobacteria. In vitro experiments with environmentally feasible concentrations (5 x 10(-7) M) of the three most common microcystins, MC-LR, -RR, and -YR, revealed that they influence the metabolism of different representative phytoplanktons. At light intensities close to the cyanobacterial bloom environment (50 mumol m(-2) s(-1)), they produce morphological and physiological changes in both microcystin-producing and nonproducing Microcystis aeruginosa strains, and also have similar effects on the green alga Scenedesmus quadricauda that is frequently present in cyanobacterial blooms. All three microcystin variants tested induce cell aggregation, increase in cell volume, and overproduction of photosynthetic pigments. All three effects appear to be related to each other, but are not necessarily caused by the same mechanism. The biological activity of microcystins toward the light-harvesting complex of photobionts can be interpreted as a signal announcing the worsening of light conditions due to the massive proliferation of cyanobacteria. Although the function of microcystins is still unknown, it is evident that they have numerous effects on phytoplankton organisms in nature. These effects depend on the individual organism as well as on the various intracellular and extracellular signaling pathways. The fact that dissolved microcystins also influence the physiology of microcystin-producing cyanobacteria leads us to the conclusion that the role of microcystins in the producing cells differs from their role in the water environment.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
33576
Microcystin-YR solution, 10 μg/mL in methanol, analytical standard
C52H72N10O13