EMAIL THIS PAGE TO A FRIEND
Medicinal research reviews
Aminopeptidase-N/CD13 (EC 3.4.11.2) inhibitors: chemistry, biological evaluations, and therapeutic prospects.
PMID 16216010
Abstract
Aminopeptidase N (APN)/CD13 (EC 3.4.11.2) is a transmembrane protease present in a wide variety of human tissues and cell types (endothelial, epithelial, fibroblast, leukocyte). APN/CD13 expression is dysregulated in inflammatory diseases and in cancers (solid and hematologic tumors). APN/CD13 serves as a receptor for coronaviruses. Natural and synthetic inhibitors of APN activity have been characterized. These inhibitors have revealed that APN is able to modulate bioactive peptide responses (pain management, vasopressin release) and to influence immune functions and major biological events (cell proliferation, secretion, invasion, angiogenesis). Therefore, inhibition of APN/CD13 may lead to the development of anti-cancer and anti-inflammatory drugs. This review provides an update on the biological and pharmacological profiles of known natural and synthetic APN inhibitors. Current status on their potential use as therapeutic agents is discussed with regard to toxicity and specificity.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
113R-1
CD13 (SP187) Rabbit Monoclonal Antibody