EMAIL THIS PAGE TO A FRIEND
Journal of endocrinological investigation
Endocrine and neuropsychological assessment in a child with a novel mutation of thyroid hormone receptor: response to 12-month triiodothyroacetic acid (TRIAC) therapy.
PMID 16218051
Abstract
To determine the endocrine, neuropsychological and genetic features of a child with resistance to thyroid hormone (RTH), and his response to long-term triiodothyroacetic acid (TRIAC) therapy. Growth, thyroid function, and neuropsychology were assessed at baseline and during 12-month TRIAC therapy. Genetic analysis was performed by PCR and denaturing high performance liquid chromatography. The main clinical finding was the attention deficit-hyperactivity disorder (ADHD). A novel mutation in exon 10 (phenylalanine to isoleucine in codon 455) was found. Long-term TRIAC therapy was effective in the management of the endocrine and neuropsychological manifestations of the syndrome. ADHD was the only phenotypic manifestation of this novel mutation of thyroid hormone (TH) receptor. TRIAC is an effective and safe drug in the long-term treatment of children with RTH.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T7650
3,3′,5-Triiodothyroacetic acid, ≥90%
C14H9I3O4