EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
Immobilization of enzymes on a microchannel surface through cross-linking polymerization.
PMID 16220171
Abstract
A novel and facile method for the preparation of an enzyme-immobilized microreactor has been developed in which enzymes are immobilized as an enzyme-polymer membrane formed on the inner wall of the microchannel by a cross-linking polymerization method; the resulting microreactor shows excellent reaction performance and stability against denaturating agents.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
G2505
N-Glutaryl-L-phenylalanine p-nitroanilide, protease substrate
C20H21N3O6