EMAIL THIS PAGE TO A FRIEND
Biotechnology letters
Efficient enzymatic synthesis of 4-methylumbelliferyl N-acetyllactosaminide and 4-methylumbelliferyl sialyl N-acetyllactosaminides employing beta-D-galactosidase and sialyltransferases.
PMID 16231217
Abstract
4-Methylumbelliferyl N-acetyllactosaminide and 4-methylumbelliferyl sialyl N-acetyllactosaminides, which are used for the assay of sialytransferase, neuraminidase and fucosyltransferase, were synthesized, respectively, by the beta-D: -galactosidase from Bacillus circulans and by a recombinant rat alpha2,3-(N)-sialyltransferase or rat liver alpha2,6-(N)-sialyltransferase with CMP-N-acetylneuraminic acid as donor.
Related Materials