EMAIL THIS PAGE TO A FRIEND
The Cochrane database of systematic reviews
Albendazole for lymphatic filariasis.
PMID 16235339
Abstract
Mass treatment with albendazole co-administered with another antifilarial drug is part of a global programme to eliminate lymphatic filariasis. We sought reliable evidence of the effects of albendazole on the disease and the parasite. To summarize the effects of albendazole alone or in combination with antifilarial drugs for clinical treatment and community control of lymphatic filariasis. We searched the Cochrane Infectious Diseases Group Specialized Register (August 2005), CENTRAL (The Cochrane Library Issue 3, 2005), MEDLINE (1966 to August 2005), EMBASE (1974 to August 2005), LILACS (1982 to August 2005), and reference lists. We also contacted researchers, the World Health Organization, and GlaxoSmithKline. Randomized and quasi-randomized controlled trials of albendazole alone or combined with another antifilarial drug for treating individuals with lymphatic filariasis, or for reducing transmission in endemic communities. Two authors independently assessed eligibility and trial quality, and extracted data. Authors contacted investigators for missing information or clarification. Seven trials including 6997 participants (995 with detectable microfilariae) met the criteria. A comparison of albendazole and placebo detected no effect on microfilariae prevalence (920 participants; 3 trials); one trial (499 participants) reported significantly lower microfilariae density at six months. Albendazole performed slightly worse than ivermectin in two trials (436 participants). Compared with diethylcarbamazine (DEC), two small trials (56 participants) found little difference in microfilariae prevalence over an extended follow up. One larger trial (502 participants) found a statistically significant effect for DEC at six months, but none at three months. Microfilariae prevalence and density were statistically significantly lower with the combination of albendazole and ivermectin compared with ivermectin alone in two of three trials (649 participants). Two trials compared albendazole plus DEC with DEC alone and found no statistically significant difference in microfilariae prevalence, though one trial favoured the combination at six months (relative risk 0.62, 95% confidence interval 0.32 to 1.21; 491 participants). This trial also found a statistically significant reduction in microfilariae density. There is insufficient evidence to confirm or refute that albendazole co-administered with DEC or ivermectin is more effective than DEC or ivermectin alone in clearing microfilariae or killing adult worms. Albendazole combined with ivermectin appears to have a small effect on microfilaraemia, but this was not consistently demonstrated. The effect of albendazole against adult and larval filarial parasites, alone and in combination with other antifilarial drugs, deserves further rigorous research.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
1012553
Albendazole, United States Pharmacopeia (USP) Reference Standard
C12H15N3O2S