EMAIL THIS PAGE TO A FRIEND
Annali di chimica
Electrocatalytical oxidation of hydroquinone with ferrocene covalently bound to L-cysteine self-assembled monolayers on a gold electrode.
PMID 16235792
Abstract
The preparation of a gold electrode modified by ferrocenecarboxylic acid (FcA) covalently bound to L-cysteine self-assembled monolayer (FcA-SAM) is described. The modified electrode shows an excellent electrocatalytic activity for the oxidation of hydroquinone (QH2) and accelerates the electron transfer rate. The anodic overpotential is reduced by ca. 290 mV compared to those obtained at bare gold electrodes. The charge transfer coefficient and the apparent surface electron transfer rate constant for the redox couple of Q/QH2 at the modified electrode are found to be 0.425 and 0.96 s(-1), respectively. The catalytic current response of DPV increases linearly with the QH2 concentration from 5.7 x 10(-7) to 3.2 x 10(-4) M. The estimation of QH2 in a simulative sample is satisfactory. The method is simple, quick, and sensitive.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
106887
Ferrocenecarboxylic acid, 97%
C11H10FeO2