EMAIL THIS PAGE TO A FRIEND
Infection and immunity
Role of lymphotoxin in experimental models of infectious diseases: potential benefits and risks of a therapeutic inhibition of the lymphotoxin-beta receptor pathway.
PMID 16239501