EMAIL THIS PAGE TO A FRIEND
Angewandte Chemie (International ed. in English)
Difluoroallenyl bromide as a wide-ranging difluoromethylene cation equivalent: SN2 substitution of difluoropropargyl bromide through sequential SE2' and SN2' reactions.
PMID 16240309