EMAIL THIS PAGE TO A FRIEND
Annali dell'Istituto superiore di sanita
Quantification of chemical elements in blood of patients affected by multiple sclerosis.
PMID 16244395
Abstract
Although some studies suggested a link between exposure to trace elements and development of multiple sclerosis (MS), clear information on their role in the aetiology of MS is still lacking. In this study the concentrations of Al, Ba, Be, Bi, Ca, Cd, Co, Cr, Cu, Fe, Hg, Li, Mg, Mn, Mo, Ni, Pb, Sb, Si, Sn, Sr, Tl, V, W, Zn and Zr were determined in the blood of 60 patients with MS and 60 controls. Quantifications were performed by inductively coupled plasma (ICP) atomic emission spectrometry and sector field ICP mass spectrometry. When the two groups were compared, an increased level of Co, Cu and Ni and a decrement of Be, Fe, Hg, Mg, Mo, Pb and Zn in blood of patients were observed. In addition, the discriminant analysis pointed out that Cu, Be, Hg, Co and Mo were able to discriminate between MS patients and controls (92.5% of cases correctly classified).
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
695092
Acetic acid, ACS reagent, ≥99.7%
C2H4O2
A6283
Acetic acid, ReagentPlus®, ≥99%
C2H4O2
338826
Acetic acid, ≥99.99% trace metals basis
C2H4O2
33209
Acetic acid, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8%
C2H4O2
27225
Acetic acid, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, 99.8-100.5%
C2H4O2
27221
Acetic acid, puriss., 99-100%
C2H4O2
45754
Acetic acid solution, for HPLC
C2H4O2
266523
Aluminum, ReagentPlus®, beads, 5-15 mm, 99.9% trace metals basis
Al
433705
Aluminum, evaporation slug, diam. × L 6.3 mm × 6.3 mm, 99.999% trace metals basis
Al
518573
Aluminum, granular, <1 mm, 99.7% trace metals basis
Al
266957
Aluminum, foil, thickness 1.0 mm, 99.999% trace metals basis
Al
266574
Aluminum, foil, thickness 0.45-0.55 mm, 99.999% trace metals basis
Al
326852
Aluminum, foil, thickness 0.25 mm, 99.999% trace metals basis
Al
326860
Aluminum, foil, thickness 0.13 mm, ≥99.99% trace metals basis
Al
11008
Aluminum, grit, ≥97.0% (complexometric)
Al
653608
Aluminum, powder, <5 μm particle size, 99.5% trace metals basis
Al
326941
Aluminum, pellets, 3-8 mesh, ≥99.999% trace metals basis
Al
338788
Aluminum, pellets, 3-12 mm, 99.99% trace metals basis
Al
202584
Aluminum, powder, <75 μm, ≥99.95% trace metals basis
Al
266515
Aluminum, ≤30 μm, spherical powder, ≥99%
Al
11009
Aluminum, powder, ≥91% (complexometric)
Al
202576
Aluminum, rod, diam. × L 3.0 mm × 100 mm, 99.999% trace metals basis
Al
310360
Aluminum, ACS reagent, 99%, wire, wire diam. ~1.5 mm
Al
266558
Aluminum, wire, diam. 1.0 mm, 99.999% trace metals basis
Al
326887
Aluminum, wire, diam. 0.58 mm, ≥99.99% trace metals basis
Al
11010
Aluminum, powder (fine), ≥93% (complexometric)
Al
IRMM523A
Aluminum, IRMM® certified Reference Material, 0.1 mm foil
Al
IRMM523B
Aluminum, IRMM® certified Reference Material, 1.0 mm foil
Al
IRMM523C
Aluminum, IRMM® certified Reference Material, 1.0 mm wire
Al
733369
Aluminum, foil, thickness 8 μm, 99% trace metals basis
Al