EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Novel histamine H3 receptor antagonists based on the 4-[(1H-imidazol-4-yl)methyl]piperidine scaffold.
PMID 16246552
Abstract
We report the discovery of novel histamine H(3) receptor antagonists based on 4-[(1H-imidazol-4-yl)methyl]piperidine. The most potent compounds in the series (e.g., 7) result from the attachment of a substituted aniline amide to the main pharmacophore piperidine via a two-methylene linker.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
255920
1,1′-(Azodicarbonyl)dipiperidine, 99%
C12H20N4O2
P16001
3-Phenoxypropionic acid, 99%
C9H10O3