EMAIL THIS PAGE TO A FRIEND
Nucleosides, nucleotides & nucleic acids
Silyl protecting groups for oligonucleotide synthesis removed by a ZnBr2 treatment.
PMID 16248081
Abstract
An oligonucleotide protected with N-(trimethylsilyloxycarbonyl) (Teoc) and P-(trimethylsilylethanol) (Tse) groups was synthesized and deprotected by a single ZnBr2 treatment. Finally it was released from the solid support by cleavage of a disulfide linkage with TCEP. The olgonucleotide was obtained without any basic treatment.