EMAIL THIS PAGE TO A FRIEND
Se pu = Chinese journal of chromatography
[Determination of praziquantel and propargite residues in fish tissues by high performance liquid chromatography with solid-phase extraction].
PMID 16250461