EMAIL THIS PAGE TO A FRIEND
The Journal of Nutrition
Reduced adiposity in bitter melon (Momordica charantia)-fed rats is associated with increased lipid oxidative enzyme activities and uncoupling protein expression.
PMID 16251604
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
AB3046 Anti-Uncoupling Protein 3 Antibody, clone, Chemicon®, from rabbit