EMAIL THIS PAGE TO A FRIEND
Nucleosides, nucleotides & nucleic acids
4-aminometyl-3-nitrobenzoic acid--a photocleavable linker for oligonucleotides containing combinatorial libraries.
PMID 16252670
Abstract
We detail the design, synthesis, and characterization of an o-nitrobenzyl-based photolabile linker containing amine and carboxyl anchor groups. A model nucleoside monomer modified with an imidazole residue and a precursor unit linked to a heterocyclic base through a photolabile tether is constructed Upon UV irradiation (313- 365 nm), the imidazole containing part of this molecule is released.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
127671
3-Aminobenzoic acid, 98%
C7H7NO2