EMAIL THIS PAGE TO A FRIEND
FEBS letters
Focal adhesion molecules as potential target of lead toxicity in NRK-52E cell line.
PMID 16253243
Abstract
In this study, we investigated the influence of inorganic lead (Pb(II)), an environmental pollutant having nephrotoxic action, on the focal adhesion (FA) organization of a rat kidney epithelial cell line (NRK-52E). In particular, we evaluated the effects of the metal on the recruitment of paxillin, focal adhesion kinase, vinculin and cytoskeleton proteins at the FAs complexes. We provided evidences that, in proliferating NRK-52E cell cultures, low concentrations of Pb(II) affect the cell adhesive ability and stimulate the disassembly of FAs, thus inhibiting the integrin-activated signalling. These effects appeared to be strictly associated to the Pb-induced arrest of cell cycle at G0/G1 phase also proved in this cell line.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
details
NRK-52E, 87012902