EMAIL THIS PAGE TO A FRIEND
Brain research. Molecular brain research
Co-expression of the 5-HT(3B) subunit with the 5-HT(3A) receptor reduces alcohol sensitivity.
PMID 16257471
Abstract
Allosteric modulation of mouse 5-Hydroxytryptamine(3A) (5-HT(3A)) and 5-HT(3A/B) receptor function by ethanol and trichloroethanol (TCEt) was assessed in HEK293 cells with whole cell patch-clamp electrophysiological recordings. Ethanol enhanced 5-HT(3A) receptor function, but had no effect on mouse 5-HT(3A/B) receptor mediated currents. The enhancing action of trichloroethanol (TCEt) on mouse 5-HT(3A/B) receptor function was much less than that observed in the mouse 5-HT(3A) receptor. Where alcohol-induced increases in peak amplitude were observed, the slope of the 20-80% rising phase of current onset was also enhanced, suggesting that increases in activation rate may be one mechanism through which alcohols enhance function of the 5-HT(3) receptors.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T54801
2,2,2-Trichloroethanol, ReagentPlus®, ≥99%
C2H3Cl3O