EMAIL THIS PAGE TO A FRIEND
The Journal of molecular diagnostics : JMD
An enhanced polymerase chain reaction assay to detect pre- and full mutation alleles of the fragile X mental retardation 1 gene.
PMID 16258159
Abstract
Several diagnostic strategies have been applied to the detection of FMR1 gene repeat expansions in fragile X syndrome. Here, we report a novel polymerase chain reaction-based strategy using the Expand Long Template PCR System (Roche Diagnostics, Mannheim, Germany) and the osmolyte betaine. Repeat expansions up to approximately 330 CGGs in males and up to at least approximately 160 CGGs in carrier women could be easily visualized on ethidium bromide agarose gels. We also demonstrated that fluorescence analysis of polymerase chain reaction products was a reliable tool to verify the presence of premutation and full mutation alleles both in males and in females. This technique, primarily designed to detect premutation alleles, can be used as a routine first screen for expanded FMR1 alleles.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B0300
Betaine solution, 5 M, PCR Reagent
C5H11NO2