EMAIL THIS PAGE TO A FRIEND
Analytical biochemistry
Multicommuted optosensor for the determination of pipemidic acid in biological fluids.
PMID 16259938
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P7903
Pipemidic acid
C14H17N5O3