EMAIL THIS PAGE TO A FRIEND
Acta physiologica Scandinavica
A single bout of exercise is followed by a prolonged decrease in the interstitial glucose concentration in skeletal muscle.
PMID 16266372
Abstract
The present study was performed to test the hypothesis that the interstitial glucose concentration in human skeletal muscle is decreased for a prolonged period following a single bout of exercise, while blood flow has returned to resting levels. Muscle interstitial concentrations of glucose, lactate, pyruvate and urea were monitored in six healthy individuals during 8 h following a 2-h one-leg exercise session by microdialysis at low perfusion flow rate. Simultaneously the blood flow was measured by the microdialysis ethanol technique. The blood glucose and the control leg interstitial glucose concentrations were stable during the experiment averaging 5.7 +/- 0.1 and 4.1 +/- 0.3 mm, respectively. In contrast, the interstitial glucose concentration in the exercise leg was markedly decreased, averaging 1.9 +/- 0.5 mm, during the first 5.5 h following exercise (P < 0.01), after which it returned towards normal values. Muscle blood flow at the site of the microdialysis catheter, measured as the ethanol outflow-to-inflow ratio, did not change significantly over time in the control or exercise leg and did not differ significantly between the two legs. Interstitial concentrations of lactate, pyruvate and urea were not significantly different between the control and exercise leg. The study shows that the interstitial glucose concentration in skeletal muscle is markedly decreased for several hours following a single exercise session. The decreased interstitial glucose concentration may serve to limit the rate of post-exercise muscle glucose uptake to a rate compatible with normal blood glucose levels and may also be speculated to have a positive long-term health implication by augmenting muscle insulin sensitivity.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
24105
Ethanol, puriss., meets analytical specification of Ph. Eur., BP, 96.0-97.2%
C2H6O
24194
Ethanol, absolute, denaturated with 0.5-1.5 Vol.% 2-butanone and approx. 0.001% Bitrex (GC), ≥98% (GC)
C2H6O
E-034
Ethanol Calibration Kit, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
C2H6O
E-040
Ethanol-10, 10 mg/dL in H2O, pack of 10 × 1.2 mL ampules, certified reference material, Cerilliant®
C2H6O
E-031
Ethanol-100 (10 ampules/kit), 100 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
C2H6O
E-038
Ethanol-100 (5 ampules/kit), 100 mg/dL in H2O, ampule of 5 × 5 mL, certified reference material, Cerilliant®
C2H6O
E-041
Ethanol-150, 150 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
C2H6O
E-056
Ethanol-20 (10 ampules/kit), 20 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
C2H6O
E-043
Ethanol-20 (5 ampules/kit), 20 mg/dL in H2O, ampule of 5 × 5 mL, certified reference material, Cerilliant®
C2H6O
E-032
Ethanol-200 (10 ampules/kit), 200 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
C2H6O
E-039
Ethanol-200 (5 ampules/kit), 200 mg/dL in H2O, ampule of 5 × 5 mL, certified reference material, Cerilliant®
C2H6O
E-035
Ethanol-25, 25 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
C2H6O
E-033
Ethanol-300, 300 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
C2H6O
E-045
Ethanol-40, 40 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
C2H6O
E-036
Ethanol-400 (10 ampules/kit), 400 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
C2H6O
E-044
Ethanol-400 (5 ampules/kit), 400 mg/dL in H2O, ampule of 5 × 5 mL, certified reference material, Cerilliant®
C2H6O
E-029
Ethanol-50, 50 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
C2H6O
E-053
Ethanol-500, 500 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
C2H6O
E-030
Ethanol-80, 80 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
C2H6O
E-037
Ethanol-80, 80 mg/dL in H2O, ampule of 5 × 5 mL, certified reference material, Cerilliant®
C2H6O
187380 Ethyl alcohol, denatured, reagent grade
459844
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
C2H6O
E7023
Ethyl alcohol, Pure, 200 proof, for molecular biology
C2H6O
493546
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
C2H6O
459828
Ethyl alcohol, Pure, 200 proof, HPLC/spectrophotometric grade
C2H6O
E7148
Ethyl alcohol, Pure, 190 proof, for molecular biology
C2H6O
270741
Reagent Alcohol, for HPLC
277649
Reagent Alcohol, anhydrous, ≤0.003% water
676829
Reagent Alcohol, anhydrous, ≤0.005% water
362808
Reagent Alcohol, reagent grade