EMAIL THIS PAGE TO A FRIEND
Organic letters
C2-symmetric bisprolinamide as a highly efficient catalyst for direct aldol reaction.
PMID 16268568
Abstract
[reaction: see text] The catalytic activity of the prolinamide-type catalysts may be improved by introducing additional prolinamide moiety into the catalyst, while the enantioselectivity can still be maintained or further improved. A C2-symmetric bisprolinamide with two prolinamide moieties has been found to be an excellent catalyst for direct aldol reaction with more than doubled reactivity and better asymmetric induction than its monoprolinamide counterpart.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
287059
L-Prolinamide, 98%
C5H10N2O