EMAIL THIS PAGE TO A FRIEND
The Journal of organic chemistry
Simple, rapid procedure for the synthesis of chloromethyl methyl ether and other chloro alkyl ethers.
PMID 16268645
Abstract
[Reaction: see text]. Zinc(II) salts catalyze the reaction between acetals and acid halides to provide haloalkyl ethers in near-quantitative yield. Reactions from millimole to mole scale are typically complete in 1-4 h with 0.01 mol % catalyst. The solutions of haloalkyl ethers thus obtained can be utilized directly in reactions in which the presence of the ester byproduct does not interfere. Excess haloalkyl ether is destroyed on workup, thereby minimizing exposure to this class of carcinogenic compounds.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
100331
Chloromethyl methyl ether, technical grade
C2H5ClO