EMAIL THIS PAGE TO A FRIEND
Chemical & pharmaceutical bulletin
Preparation of (Cyanomethylene)tributylphosphorane: a new mitsunobu-type reagent.
PMID 16272744
Abstract
(Cyanomethylene)tributylphosphorane (CMBP), which promoted the alkylation of various nucleophiles (HA) with alcohols (ROH) to give RA (Mitsunobu-type reaction), was prepared in two steps starting from chloroacetonitrile.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C19651
Chloroacetonitrile, 99%
C2H2ClN