EMAIL THIS PAGE TO A FRIEND
International journal of oncology
Cytotoxicity of the polyamine oxidase inactivator MDL 72527 to cancer cells: comparison with a saturated structural analogue.
PMID 16273223
Abstract
MDL 72527 (N1,N4-di-2,3-butadienyl-1,4-butanediamine) is a selective inactivator of polyamine oxidase with therapeutic potential. However, the development of lethal toxic effects due to prevention of spermine degradation is a considerable disadvantage of the compound. Since the cytotoxicity of MDL 72527 was postulated to be independent of its anti-polyamine oxidase activity, its cytotoxicity to cancer cells was compared with that of a close analogue that is devoid of structural features enabling mechanism-based inactivation of polyamine oxidase. N1,N4-di-n-butyl-1,4-butanediamine proved to be a cytotoxic agent of considerable potency, which induces mainly non-apoptotic cell death, whereas MDL 72527 causes under identical conditions both, apoptotic and non-apoptotic cell death. The sensitivity of cells to both compounds is presumably dependent of their glutathione content.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M2949
MDL 72527, ≥98% (HPLC)
C12 H20 N2·2 HCl