EMAIL THIS PAGE TO A FRIEND
Biotechnology and bioengineering
Prevention of thermal induced aggregation of cytochrome c at isoelectric pH values by polyanions.
PMID 16273554
Abstract
Differential scanning calorimetry, viscometry, optical and CD spectroscopy were used to characterize the influence of two polyanions, poly(vinylsulfate) (PVS), and poly(4-styrene-sulfonate) (PSS) on thermal transition reversibility of ferricytochrome c at or near isoelectric pH. In these conditions, both PVS and PSS enhance the thermal transition reversibility of cytochrome c by preventing the aggregation of denatured protein molecules. Data indicate that the polyanions are in complex with cytochrome c that is stabilized by synergistic effect of Coulombic and non-Coulombic interactions.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
271969
Poly(vinyl sulfate) potassium salt, average Mw ~170,000