EMAIL THIS PAGE TO A FRIEND
World journal of gastroenterology
Role of nuclear receptor CAR in carbon tetrachloride-induced hepatotoxicity.
PMID 16273607
Abstract
To investigate the precise roles of CAR in CCl(4)-induced acute hepatotoxicity. To prepare an acute liver injury model, CCl(4) was intraperitoneally injected in CAR+/+ and CAR-/- mice. Elevation of serum alanine aminotransferase and extension of centrilobular necrosis were slightly inhibited in CAR-/- mice compared to CAR+/+ mice without PB. Administration of a CAR inducer, PB, revealed that CCl(4)-induced liver toxicity was partially inhibited in CAR-/- mice compared with CAR+/+ mice. On the other hand, androstanol, an inverse agonist ligand, inhibited hepatotoxicity in CAR+/+ but not in CAR-/- mice. Thus, CAR activation caused CCl(4) hepatotoxicity while CAR inhibition resulted in partial protection against CCl(4)-induced hepatotoxicity. There were no differences in the expression of CYP2E1, the main metabolizing enzyme for CCl(4), between CAR+/+ and CAR-/- mice. However, the expression of other CCl(4)-metabolizing enzymes, such as CYP2B10 and 3A11, was induced by PB in CAR+/+ but not in CAR-/- mice. Although the main pathway of CCl(4)-induced acute liver injury is mediated by CYP2E1, CAR modulates its pathway via induction of CYP2B10 and 3A11 in the presence of activator or inhibitor. The nuclear receptor CAR modulates CCl(4)-induced liver injury via induction of CCl(4)-metabolizing enzymes in the presence of an activator. Our results suggest that drugs interacting with nuclear receptors such as PB might play critical roles in drug-induced liver injury or drug-drug interaction even though such drugs themselves are not hepatotoxic.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A2480
5α-Androstan-3β-ol, powder
C19H32O